missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide-d7, for NMR, 99.5 atom % D

Product Code. 16388186 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
Unit Size:
10mL
1mL
25mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
16388186 25 mL Glass Bottle 25mL
10479670 1 mL Glass Bottle 1mL
16378186 5 mL Glass Bottle 5mL
10762611 10 mL Glass Bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16388186 Supplier Thermo Scientific Acros Supplier No. 183600250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4472-41-7 | C3H7NO | 80.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4472-41-7
Molecular Formula C3H7NO
MDL Number MFCD00003286
InChI Key ZMXDDKWLCZADIW-YYWVXINBSA-N
Synonym dimethylformamide d, formamide-1-d, n,n-di methyl-d3, deuterated dmf, n,n-dimethylformamide-d7, n,n-dimethylformamide-d7 99.5atom%d, n,n-dimethylformamide-d7 dmf-d7, dimethyl formamide-d7, heptadeutero-n,n-dimethylformamide, dmf-d7, n,n-di 2h3 methyl 2h formamide
PubChem CID 78225

Specifications

Physical Form Liquid
Color Colorless
Density 1.0300 g/mL
Deuterium (D) 99.4% min.
Flash Point 58°C
Packaging Glass Bottle
Refractive Index 1.428
Water 0.1% max. (Coulometric)
Quantity 25 mL
Linear Formula DCON(CD3)2
SMILES [2H]C(=O)N(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name N,N-di(²H₃)methyl(²H)formamide
Molecular Weight (g/mol) 80.14
Formula Weight 80.15
Grade NMR
Chemical Name or Material N, N-Dimethylformamide-d7, 99.5 atom % D
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.