missing translation for 'onlineSavingsMsg'
Learn More

Solstice N,N-Dimethylformamide CHROMASOLV™

Product Code. 16605843
Change view
Click to view available options
Quantity:
18 L
Unit Size:
18 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16605843 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16605843 Supplier Solstice Supplier No. 3469918L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N,N-Dimethylformamide CHROMASOLV™ for HPLC, ≥99.9% (GC).

  • ≥99.9% (GC)

Specifications

pH 7.0 (20°C)
Melting Point -61°C
CAS 68-12-2
CAS Min % ≥90.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.94 g/cm3 (20°C)
Quantity 18 L
Flash Point 57.5°C (1,013.25 hPa)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line CHROMASOLV
Molecular Formula C3H7NO
Vapor Pressure 23 hPa (50°C)
MDL Number MFCD00003284
Viscosity 0.86 mPa.s (20°C)
UN Number UN2265
Beilstein 605365
Synonym DMF, Diemethyl formamide
Boiling Point 152°C to 153°C (1,013 hPa)
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES CN(C)C=O
IUPAC Name N,N-dimethylformamide
Molecular Weight (g/mol) 73.10
Percent Purity ≥99.90%
Grade HPLC
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.