missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, anhydrous, amine free, 99.9%

Product Code. p-4408416
Click to view available options
Quantity:
4 L
4 x 4 L
Unit Size:
16 L
4 L
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11315787

Brand: Thermo Scientific Alfa Aesar 043465.K7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Suitable For Nucleic Acids, Peptide Synthesis | CAS: 68-12-2 | C3H7NO | 73.10 g/mol

Suitable for nucleic acids, peptide synthesisN, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Melting Point -61°C
Color Colorless
Density 0.944 g/cm3 at 20°C
Boiling Point 153°C
Flash Point 57°C (135°F)
Ignition Point 440°C (824°F)
Assay Percent Range 99.9%
Odor Amine-like
Refractive Index 1.431
Quantity 4 L
UN Number 2265
Beilstein 605365
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Vapor Pressure 3mm Hg at 20°C
Physical Form Liquid
Chemical Name or Material N, N-Dimethylformamide, Contains amine and water scavenging sieve
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.