missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99.8%, for spectroscopy

Product Code. 11369716 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11369716 1 L Glass bottle 1 L
10533151 500 mL Glass bottle 500mL
10154180 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11369716 Supplier Thermo Scientific Acros Supplier No. 167790010
  • ONLINE EXCLUSIVE PRICE
    £50.10
    £48.60 / 1 L
    Save £1.50
    2% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 68-12-2 | C3H7NO | 73.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-12-2
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00003284
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
PubChem CID 6228
ChEBI CHEBI:17741
IUPAC Name N,N-dimethylformamide
SMILES CN(C)C=O

Specifications

Alkalinity 0.001 meq/g max.
Acidity 0.001 meq/g max.
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 340nm, 0.05 max. at 310nm, 0.1 max. at 295nm, 0.3 max. at 275nm, 1 max. at 270nm
CAS Min % 99.75
CAS Max % 100.0
Melting Point -61.0°C
Density 0.9450g/mL
Boiling Point 153.0°C
Flash Point 58°C
Assay Percent Range 99.75% min. (GC)
Packaging Glass bottle
Linear Formula HCON(CH3)2
Refractive Index 1.4290 to 1.4320
Quantity 1 L
Beilstein 04, 58
Residue after Evaporation 0.0015% max.
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Specific Gravity 0.945
Specific Rotation 0.94
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Viscosity 0.8 mPa.s (20°C)
Formula Weight 73.09
Percent Purity 99.8%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.