missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99.8%, Extra Dry, AcroSeal™, ACROS Organics™

Brand:  Acros Organics 326870025

Code : 29

Additional Details : CAS Number : 68-12-2 Weight : 3.85000kg Transport : UN number : 2265 Chem class : 1 Pack group : III

Quantity 2.5L
Packaging Glass bottle

 View more versions of this product

Product Code. 10437392

  • £147.50 / 2.50 L


EU Stock 39
Add to basket

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Specifications

Chemical Identifiers

N,N-Dimethylformamide
99.75
Extra Dry
99.8%
C3H7NO
73.09
DMF
68-12-2
100.0
Water <50ppm
99.8%
HCON(CH3)2
MFCD00003284

Specifications

N,N-Dimethylformamide
99.75
0.002 % max.
Water <50ppm
99.8%
C3H7NO
73.09
04, 58
15, 3267
Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Authentic
1.4290 to 1.4320
Solubility in water: soluble. Other solubilities: miscible with most organic solvents
0.94
58°C
153.0°C
− 61.0°C
68-12-2
100.0
Extra Dry
Liquid
99.8%
HCON(CH3)2
MFCD00003284
01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
DMF
0.9450g/mL
Glass bottle
0.001 % max.
0.945
0.005% Max. (Coulometric)
0.8 mPa.s (20°C)
Clear colorless
2.5L
Videos
SDS Documentation

Technical Documents