missing translation for 'onlineSavingsMsg'
Learn More

Solstice N,N-Dimethylformamide

Product Code. 16625743 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
190 kg
Unit Size:
190kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16625743 190 kg 190kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16625743 Supplier Solstice Supplier No. 15440S190KG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purum

  • Purum

Specifications

pH 7.0 (20°C)
Melting Point -61°C
CAS 68-12-2
CAS Min % ≥90.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.94 g/cm3 (20°C)
Quantity 190 kg
Flash Point 57.5°C (1.013.25 hPa)
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Purum
Molecular Formula C3H7NO
Vapor Pressure 3.77 hPa at 20°C (68°F), 23 hPa at 50°C (122°F)
MDL Number MFCD00003284
Viscosity 0.86 mPa.s (20°C)
UN Number UN2265
Beilstein 605365
Concentration ≤100.00%
Synonym DMF, Diemethyl formamide
Boiling Point 152°C to 153°C (1,013 hPa)
InChI Key ZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILES CN(C)C=O
IUPAC Name N,N-dimethylformamide
Molecular Weight (g/mol) 73.10
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.