missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyldodecylamine-d6, TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyldodecylamine-d6 |
| Molecular Formula | C14 D6 H25 N |
| Quantity | 100 mg |
| Synonym | (Dimethylamino)dodecane-d6, 1-Dimethylaminododecane-d6, Adma 12-d6, Antioxidant DDA-d6, Armeen DM 12D-d6, Barlene 12S-d6, DDA-d6, DDA (corrosion inhibitor)-d6, DM 2098-d6, Dodecyldimethylamine-d6, Empigen AB-d6, Farmin DM 20-d6, Farmin DM 2098-d6, Genamin LA-d6, 302D-d6, IPL 12-d6, Lauryldimethylamine-d6, Monolauryl Dimethylamine-d6, N,N-Dimethyl-1-dodecanamine-d6, N,N-Dimethyl-n-dodecylamine-d6, N,N-Dimethyllaurylamine-d6, N-Dodecyl-N,N-dimethylamine-d6, N-Dodecyldimethylamine-d6, N-Lauryldimethylamine-d6, NSC 7332-d6, Nissan Tertiary Amine BB-d6, RC 5629-d6, Toyocat D 60-d6 |
| InChI Formula | InChI=1S/C14H31N/c1-4-5-6-7-8-9-10-11-12-13-14-15(2)3/h4-14H2,1-3H3/i2D3,3D3 |
| SMILES | [2H]C([2H])([2H])N(CCCCCCCCCCCC)C([2H])([2H])[2H] |
| IUPAC Name | N,N-bis(trideuteriomethyl)dodecan-1-amine |
| Molecular Weight (g/mol) | 219.44 |
| Formula Weight | 219.283 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?