missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldodecylamine, 90+%

Product Code. 11481157
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481157 250 g 250g
11471157 50 g 50g
11461157 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481157 Supplier Thermo Scientific Alfa Aesar Supplier No. B24145.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 112-18-5 | C14H31N | 213.409 g/mol

N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyldodecylamine is used as a as a capping agent during the synthesis of Au(core) -Pd(shell) bimetallic nanoparticles in toluene. It is used in the synthesis of N-alkyl-N,N-dimethyl-1-ammonio-2-hydroxy-3-propane sulfonate. Also used in the preparation of novel bis-quaternary ammonium salt.

Solubility
Slightly soluble in water.

Notes
Air & moisture sensitive. Materials to avoid is copper, oxidizing agents, strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-18-5
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.409
MDL Number MFCD00008970
InChI Key YWFWDNVOPHGWMX-UHFFFAOYSA-N
Synonym n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
PubChem CID 8168
IUPAC Name N,N-dimethyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)C

Specifications

Melting Point -20°C
Density 0.787
Boiling Point 110°C to 112°C (3 mmHg)
Flash Point 118°C (244°F)
Refractive Index 1.4375
Quantity 250 g
UN Number UN2735
Sensitivity Air and moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 213.41
Percent Purity ≥90%
Chemical Name or Material N,N-Dimethyldodecylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.