missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyldecanamide, 98%

Product Code. 11322686
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322686 25 g 25g
11312686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322686 Supplier Thermo Scientific Alfa Aesar Supplier No. L08731.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14433-76-2 | C12H25NO | 199.338 g/mol

N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14433-76-2
Molecular Formula C12H25NO
Molecular Weight (g/mol) 199.338
MDL Number MFCD00043725
InChI Key HNXNKTMIVROLTK-UHFFFAOYSA-N
Synonym decanamide, n,n-dimethyl, n,n-dimethylcapramide, n,n-dimethylcapylamide, n,n-dimethyldecanoamide, unii-o29y6x2jez, n,n-dimethyldecan-1-amide, nn-dimethyldecanamide, n,n-dimethyl decanamide, o29y6x2jez, dimethyl capramide
PubChem CID 26690
IUPAC Name N,N-dimethyldecanamide
SMILES CCCCCCCCCC(=O)N(C)C

Specifications

Boiling Point 110°C to 111°C (0.5 mmHg)
Refractive Index 1.454
Quantity 25 g
Beilstein 1906042
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 199.34
Percent Purity 98%
Chemical Name or Material N,N-Dimethyldecanamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.