missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylcyclohexylamine, 98+%

Product Code. 11363566
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363566 100 mL 100mL
11353566 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363566 Supplier Thermo Scientific Alfa Aesar Supplier No. L14521.AE
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-94-2 | C8H17N | 127.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-94-2
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00003844
InChI Key SVYKKECYCPFKGB-UHFFFAOYSA-N
Synonym n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl
PubChem CID 7415
ChEBI CHEBI:59022
IUPAC Name N,N-dimethylcyclohexanamine
SMILES CN(C)C1CCCCC1

Specifications

Melting Point -60°C
Density 0.849
Boiling Point 160°C to 161°C
Flash Point 39°C (102°F)
Refractive Index 1.454
Quantity 100 mL
UN Number UN2264
Beilstein 1919922
Formula Weight 127.23
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethylcyclohexylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.