missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzeneethanamine Hydrochloride, TRC

Product Code. 30387646
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387646 10 g -
30374908 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387646 Supplier Toronto Research Chemicals Supplier No. TRCD46132510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10275-21-5
Molecular Formula C10 H15 N . Cl H
Molecular Weight (g/mol) 185.6937
Synonym N,N-DMPEA
IUPAC Name N,N-dimethyl-2-phenylethanamine;hydrochloride
SMILES Cl.CN(C)CCc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H15N.ClH/c1-11(2)9-8-10-6-4-3-5-7-10;/h3-7H,8-9H2,1-2H3;1H
Formula Weight 185.0971
Chemical Name or Material N,N-Dimethylphenethylamine (hydrochloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.