missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline, 99%

Product Code. 10356600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
10 L
2.5 L
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356600 5 mL Glass Bottle 5mL
10336650 1 L Glass Bottle 1 L
10580571 2.5 L Glass Bottle 2.5 L
10125633 10 L Metal drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10356600 Supplier Thermo Scientific Acros Supplier No. 115920050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-69-7 | C8H11N | 121.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008304
InChI Key JLTDJTHDQAWBAV-UHFFFAOYSA-N
Synonym dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
PubChem CID 949
ChEBI CHEBI:16269
IUPAC Name N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 1.5°C to 2.5°C
Color Yellow
Density 0.9500g/mL
Boiling Point 193.0°C to 194.0°C
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula C6H5N(CH3)2
Refractive Index 1.5570 to 1.5590
Quantity 5 mL
Fieser 01,274; 02,147
Merck Index 15, 3256
Specific Gravity 0.95
Solubility Information Solubility in water: 1g/L (20°C). Other solubilities: freely soluble in alcohol, ether, chloroform, acet, carbon tetrachloride
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.