missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylaniline, 99%

Product Code. 11488770
Change view
Click to view available options
Quantity:
10,000 mL
100 mL
2500 mL
500 mL
Unit Size:
10000mL
100mL
2500mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488770 100 mL 100mL
11483894 500 mL 500mL
11493894 2500 mL 2500mL
11403904 10,000 mL 10000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11488770 Supplier Thermo Scientific Alfa Aesar Supplier No. A11916.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-69-7 | C8H11N | 121.18 g/mol

N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler's ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylaniline acts as a precursor to triarylmethane dyes such as crystal violet and malachite green. It is also used in the synthesis of vanillin, methyl violet, and Michler′s ketone. Further, it is used as a hardener for plastic resins and an acid scavenger in the manufacture of semisynthetic penicillins and cephalosporins. In addition to this, it is used as a promoter in the curing of polyester and vinyl ester resins.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, benzene and alkaline.

Notes
Incompatible with strong oxidizing agents, Strong acids, acid chlorides, acid anhydrides, chloroformates and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-69-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008304
InChI Key JLTDJTHDQAWBAV-UHFFFAOYSA-N
Synonym dimethylaniline, dimethylphenylamine, n,n-dimethylbenzenamine, benzenamine, n,n-dimethyl, dimethylamino benzene, n,n-dimethylphenylamine, n,n-dimethylbenzeneamine, dimethylaminobenzene, dwumetyloanilina, n,n-dimethylamino benzene
PubChem CID 949
ChEBI CHEBI:16269
IUPAC Name N,N-dimethylaniline
SMILES CN(C)C1=CC=CC=C1

Specifications

Melting Point 2°C
Density 0.956
pH 7.4
Boiling Point 193°C
Flash Point 62°C (143°F)
Assay Percent Range 99%
Odor Characteristic
Refractive Index 1.558
Quantity 100 mL
UN Number UN2253
Beilstein 507140
Merck Index 14,3234
Solubility Information Miscible with water,ethanol,ether,acetone,chloroform,benzene and alkaline.
Formula Weight 121.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H351-Suspected of causing cancer.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: N,N-Dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.