missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacrylamide, 99%, stabilized with 500 ppm MEHQ

Product Code. 11379671 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11379671 100 mL Glass bottle 100mL
11915481 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379671 Supplier Thermo Scientific Acros Supplier No. 432021000
  • ONLINE EXCLUSIVE PRICE
    £45.40
    £37.65 / 100mL
    Save £7.75
    17% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2680-03-7 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2680-03-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
InChI Key YLGYACDQVQQZSW-UHFFFAOYSA-N
Synonym n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
PubChem CID 17587
IUPAC Name N,N-dimethylprop-2-enamide
SMILES CN(C)C(=O)C=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9620g/mL
Boiling Point 80.0°C (20.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4720 to 1.4740 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 0.962
Solubility Information Solubility in water: slightly soluble.
Stabilizer 500ppm (approximately)
Formula Weight 99.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylacrylamide, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacrylamide, stabilized with 500 ppm MEHQ
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.