missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol

Product Code. 11384548
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384548 50 g 50g
11394548 250 g 250g
11304558 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384548 Supplier Thermo Scientific Alfa Aesar Supplier No. 042405.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2680-03-7 | C5H9NO | 99.13 g/mol

N,N-Dimethylacrylamide is a useful monomer for synthetic fibers and plastics. It is also used in the synthesis of photoresponsive polymers and copolymers by grafting polysiloxane on the side chain of DMA by homopolymerization or grafting cellulose to form cellulose polymers and copolymerization of methyl methacrylate with DMA or copolymerization of DMA with styrene and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2680-03-7
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00008626
InChI Key YLGYACDQVQQZSW-UHFFFAOYSA-N
Synonym n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
PubChem CID 17587
IUPAC Name N,N-dimethylprop-2-enamide
SMILES CN(C)C(=O)C=C

Specifications

Color Colorless
Density 0.962 g/mL
Boiling Point 82°C to 84°C (21 mmHg)
Flash Point 71°C (161°F)
Linear Formula H2C=CHCON(CH3)2
Refractive Index 1.473
Quantity 50 g
UN Number UN2810
Beilstein 1742219
Sensitivity Light sensitive
Solubility Information Slightly soluble in water (1.000g/L at 20°C).
Formula Weight 99.13
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylacrylamide, Stabilized with 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.