missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide-d{9}, 99% (Isotopic), Thermo Scientific Chemicals

Catalog Number 11350528
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
116057-81-9
C4H9NO
87.12
MFCD00066061
FXHOOIRPVKKKFG-UHFFFAOYSA-N
n,n-dimethylacetamide-d9, n,n-bis ∼2∼h_3_ methyl ∼2∼h_3_ acetamide, an,n-dimethylacetamide-d9, n,n-dimethylacetamide-d9, 99 atom % d, n,n-bis 2 h? methyl 2 h? acetamide, n,n-di 2 h? methyl 2 h? acetamide
22789542
CN(C)C(C)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
116057-81-9
C4H9NO
87.12
MFCD00066061
FXHOOIRPVKKKFG-UHFFFAOYSA-N
n,n-dimethylacetamide-d9, n,n-bis ∼2∼h_3_ methyl ∼2∼h_3_ acetamide, an,n-dimethylacetamide-d9, n,n-dimethylacetamide-d9, 99 atom % d, n,n-bis 2 h? methyl 2 h? acetamide, n,n-di 2 h? methyl 2 h? acetamide
22789542
CN(C)C(C)=O

N,N-Dimethylacetamide-d{9} is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.035 g/mL
Flash Point 70°C (158°F)
Packaging Ampule
Refractive Index 1.438
Quantity 1 g
Melting Point -20°C
Boiling Point 165°C
Linear Formula CD3CON(CD3)2
Solubility Information Miscible with water.
IUPAC Name 2,2,2-trideuterio-N,N-bis(trideuteriomethyl)acetamide
Formula Weight 96.19
Percent Purity 99%
Chemical Name or Material N,N-Dimethylacetamide-d9, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'