missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide-d{9}, 99% (Isotopic)

Product Code. 11340528
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340528 5 g 5g
11350528 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11340528 Supplier Thermo Scientific Alfa Aesar Supplier No. 043854.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 116057-81-9 | C4H9NO | 87.12 g/mol

N,N-Dimethylacetamide-d{9} is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116057-81-9
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00066061
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym n,n-dimethylacetamide-d9, n,n-bis ∼2∼h_3_ methyl ∼2∼h_3_ acetamide, an,n-dimethylacetamide-d9, n,n-dimethylacetamide-d9, 99 atom % d, n,n-bis 2 h? methyl 2 h? acetamide, n,n-di 2 h? methyl 2 h? acetamide
PubChem CID 22789542
SMILES CN(C)C(C)=O

Specifications

Physical Form Liquid
Color Colorless
Density 1.035 g/mL
Flash Point 70°C (158°F)
Packaging Ampule
Refractive Index 1.438
Quantity 5 g
Melting Point -20°C
Boiling Point 165°C
Linear Formula CD3CON(CD3)2
Solubility Information Miscible with water.
IUPAC Name N,N-dimethylacetamide
Formula Weight 96.19
Percent Purity 99%
Chemical Name or Material N,N-Dimethylacetamide-d9, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.