missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, >99%, MP Biomedicals™

Product Code. 11486692 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486692 250 mL 250mL
11496692 500 mL 500mL
11406702 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11486692 Supplier MP Biomedicals Supplier No. 0215092883
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 127-19-5
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00008686
InChI Key FXHOOIRPVKKKFG-UHFFFAOYSA-N
Synonym dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
PubChem CID 31374
ChEBI CHEBI:84254
IUPAC Name N,N-dimethylacetamide
SMILES CN(C)C(C)=O

Specifications

Color Colorless
Density 0.94g/cm3 at 20°C
Quantity 250 mL
Physical Form Liquid
Assay Percent Range >99%
Formula Weight 87.12
Chemical Name or Material N, N-Dimethylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.