missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Cytokeratin 14 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | KRT14/7054R |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ |
| Host Species | Rabbit |
| Immunogen | Recombinant human Cytokeratin 14 fragment (around aa350-472) (exact sequence is proprietary) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?