missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-toluidine, 99%

Product Code. 11414034
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414034 500 g 500g
11404034 100 g 100g
11424034 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414034 Supplier Thermo Scientific Alfa Aesar Supplier No. A11988.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99-97-8 | C9H13N | 135.21 g/mol

N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-97-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008316
InChI Key GYVGXEWAOAAJEU-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
PubChem CID 7471
IUPAC Name N,N,4-trimethylaniline
SMILES CN(C)C1=CC=C(C)C=C1

Specifications

Melting Point -25°C
Density 0.937
Boiling Point 210°C to 211°C
Flash Point 83°C (181°F)
Refractive Index 1.546
Quantity 500 g
UN Number UN1708
Beilstein 774409
Solubility Information Miscible with alcohol,ether and chloroform. Immiscible with water.
Formula Weight 135.21
Percent Purity 99%
Chemical Name or Material N,N-Dimethyl-p-toluidine
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.