missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-toluidine, 99%, Thermo Scientific Chemicals

Catalog Number 11404034
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
99-97-8
C9H13N
135.21
MFCD00008316
GYVGXEWAOAAJEU-UHFFFAOYSA-N
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
7471
N,N,4-trimethylaniline
CN(C)C1=CC=C(C)C=C1
This item is not returnable. View return policy
99-97-8
C9H13N
135.21
MFCD00008316
GYVGXEWAOAAJEU-UHFFFAOYSA-N
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
7471
N,N,4-trimethylaniline
CN(C)C1=CC=C(C)C=C1

N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-toluidine is used as a polymerization catalyst for polyesters, acrylate and epoxy resins. It is also used as a hardener for dental cements and in adhesives. It serves as an intermediate for photographic chemicals, in industrial glues, in artificial fingernail preparations, colorants, pharmaceuticals. It reacts with vinyl ether in the presence of copper(II) chloride gives tetrahydroquinolines. Further, it is used to accelerate polymerization of ethyl methacrylate.

Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents. Store in a cool place.
TRUSTED_SUSTAINABILITY
Melting Point -25°C
Density 0.937
Boiling Point 210°C to 211°C
Flash Point 83°C (181°F)
Refractive Index 1.546
Quantity 100 g
UN Number UN1708
Beilstein 774409
Solubility Information Miscible with alcohol,ether and chloroform. Immiscible with water.
Formula Weight 135.21
Percent Purity 99%
Chemical Name or Material N,N-Dimethyl-p-toluidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'