missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine-d6, TRC

Product Code. 30382937
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30382937 100 mg
30378986 25 mg
30370292 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382937 Supplier Toronto Research Chemicals Supplier No. TRCD476762100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1398066-19-7
Molecular Formula C8 D6 H6 N2
Molecular Weight (g/mol) 142.231
Synonym Dimethyl-p-phenylenediamine-d6, N,N-Di(methyl-d3)-1,4-phenylenediamine, N1,N1-Di(methyl-d3)-1,4-benzenediamine, N,N-Dimethyl-p-phenylenediamine-D6 (N,N-dimethyl-D6), N,N-Dimethyl-p-phenylenediamine-d6, 4-Amino-N,N-dimethyl-d6-aniline
IUPAC Name 1-N,1-N-bis(trideuteriomethyl)benzene-1,4-diamine
SMILES [2H]C([2H])([2H])N(c1ccc(N)cc1)C([2H])([2H])[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H12N2/c1-10(2)8-5-3-7(9)4-6-8/h3-6H,9H2,1-2H3/i1D3,2D3
Formula Weight 142.138
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.