missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-phenylenediamine, 96%

Product Code. 11488526
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488526 250 g 250g
11478526 50 g 50g
11468526 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488526 Supplier Thermo Scientific Alfa Aesar Supplier No. A15962.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 99-98-9 | C8H12N2 | 136.20 g/mol

N,N-Dimethyl-p-phenylenediamine is a redox indicator. Readily forms stable, red radical cation, involved in a variety of redox reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-p-phenylenediamine is a redox indicator. Readily forms stable, red radical cation, involved in a variety of redox reactions.

Solubility
Soluble in Water (11 g/L at20°C), diethyl ether, petroleum ether, alcohol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light and air sensitive. Store under inert gas. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-98-9
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.20
MDL Number MFCD00007860
InChI Key BZORFPDSXLZWJF-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-phenylenediamine, n,n-dimethyl-1,4-phenylenediamine, n1,n1-dimethylbenzene-1,4-diamine, p-aminodimethylaniline, dmpd, 4-dimethylamino aniline, dimethyl-p-phenylenediamine, p-amino-n,n-dimethylaniline, 4-amino-n,n-dimethylaniline, p-dimethylamino aniline
PubChem CID 7472
ChEBI CHEBI:15783
IUPAC Name 4-N,4-N-dimethylbenzene-1,4-diamine
SMILES CN(C)C1=CC=C(N)C=C1

Specifications

Melting Point 34°C to 38°C
Density 1.09
Boiling Point 262°C
Flash Point 130°C (266°F)
Quantity 250 g
UN Number UN2811
Beilstein 508105
Merck Index 14,3254
Solubility Information Soluble in Water (11g/L at20°C),diethyl ether,petroleum ether,alcohol and chloroform.
Formula Weight 136.2
Percent Purity 96%
Chemical Name or Material N,N-Dimethyl-p-phenylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.