missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-n-octylamine, 95%

Product Code. 11474234
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474234 250 g 250g
11454234 10 g 10g
11464234 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11474234 Supplier Thermo Scientific Alfa Aesar Supplier No. A12546.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7378-99-6 | C10H23N | 157.30 g/mol

N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-n-octylamine is used as ion-pairing agent in the quantitative estimation of thiazinamium methylsulphate in a pharmaceutical preparation by HPLC. It acts as a counterion in mobile phase for separation of tetracycline, tetracycline analogs and their potential impurities by reversed-phase ion-pair chromatography. Further, it is used in the study of chiral separations by complexation with proteins in capillary zone electrophoresis. In addition to this, it is used as a mobile phase in direct resolution and quantitation of (R)- and (S)-disopyramide

Solubility
Slightly miscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents, strong acids and copper.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7378-99-6
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00009558
InChI Key UQKAOOAFEFCDGT-UHFFFAOYSA-N
Synonym n,n-dimethyloctylamine, n,n-dimethyl-n-octylamine, dimethyloctylamine, octyldimethylamine, dimethyl octyl amine, n-octyldimethylamine, dimethyl-n-octylamine, 1-octanamine, n,n-dimethyl, octylamine, n,n-dimethyl, unii-20n7h7x4sd
PubChem CID 16224
IUPAC Name N,N-dimethyloctan-1-amine
SMILES CCCCCCCCN(C)C

Specifications

Melting Point -57°C
Density 0.765
Boiling Point 195°C
Flash Point 65°C (149°F)
Refractive Index 1.4243
Quantity 250 g
UN Number UN2922
Beilstein 1698081
Sensitivity Air Sensitive
Solubility Information Slightly miscible with water.
Formula Weight 157.3
Percent Purity 95%
Chemical Name or Material N,N-Dimethyl-n-octylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.