missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-toluidine, 97+%

Product Code. 11480933
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480933 5 g 5g
11490933 25 g 25g
11400943 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480933 Supplier Thermo Scientific Alfa Aesar Supplier No. A13326.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-72-2 | C9H13N | 135.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-72-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00008305
InChI Key CWOMTHDOJCARBY-UHFFFAOYSA-N
Synonym n,n-dimethyl-m-toluidine, benzenamine, n,n,3-trimethyl, dimethyl-m-toluidine, m-methyl-n,n-dimethylaniline, n,n-dimethyl-m-methylaniline, m,n,n-trimethylaniline, n,n-dimethyl-3-methylaniline, dimetil-m-toluidina, n,n,3-trimethylbenzenamine, benzene, 1-dimethylamino-3-methyl
PubChem CID 8488
IUPAC Name N,N,3-trimethylaniline
SMILES CN(C)C1=CC=CC(C)=C1

Specifications

Melting Point -15°C
Boiling Point 212°C to 215°C
Flash Point 85°C (185°F)
Quantity 5 g
UN Number UN1708
Beilstein 1422766
Formula Weight 135.21
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-m-toluidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.