missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%

Product Code. 11340024
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340024 5 g 5g
11330024 1 g 1g
11350024 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340024 Supplier Thermo Scientific Alfa Aesar Supplier No. L00403.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3575-32-4 | C8H14Cl2N2 | 209.114 g/mol

it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
it can be used to produce N-(4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-yl)-formamide with 4,5,6-trimethoxy-2,4-dimethyl-tetrahydro-pyran-3-ylamine. This reaction will need solvent tetrahydrofuran. it is applied as a selective formylating agent for one of two amino groups in such basic amino acids as ornithine or lysine.

Solubility
Soluble in water, chloroform and dichloromethane. Slightly soluble in ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3575-32-4
Molecular Formula C8H14Cl2N2
Molecular Weight (g/mol) 209.114
MDL Number MFCD00012971
InChI Key BZJPIQKDEGXVFG-UHFFFAOYSA-N
Synonym n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
PubChem CID 77124
IUPAC Name 3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
SMILES CN(C)C1=CC=CC(=C1)N.Cl.Cl

Specifications

Melting Point 216°C (decomposition)
Quantity 5 g
UN Number UN2811
Beilstein 3694384
Solubility Information Soluble in water,chloroform and dichloromethane. Slightly soluble in ethyl acetate.
Formula Weight 209.12
Percent Purity 99%
Chemical Name or Material N,N-Dimethyl-m-phenylenediamine dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.