missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-m-phenylenediamine dihydrochloride, 99%

Product Code. 10351514 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351514 100 g Glass bottle 100g
10791091 5 g Glass bottle 5g
10368050 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351514 Supplier Thermo Scientific Acros Supplier No. 204341000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 3575-32-4 | C8H12N2·2HCl | 209.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3575-32-4
Molecular Formula C8H12N2·2HCl
Molecular Weight (g/mol) 209.12
MDL Number MFCD00012971
InChI Key BZJPIQKDEGXVFG-UHFFFAOYSA-N
Synonym n1,n1-dimethylbenzene-1,3-diamine dihydrochloride, n,n-dimethyl-m-phenylenediamine dihydrochloride, m-dimethylamino aniline, n,n-dimethyl-1,3-phenylenediamine dihydrochloride, 1,3-benzenediamine, n,n-dimethyl-, dihydrochloride, n,n-dimethylbenzene-1,3-diamine dihydrochloride, 3-amino-n,n-dimethylaniline, 1,3-benzenediamine, n1,n1-dimethyl-, hydrochloride 1:2, pubchem14991, acmc-20a5k9
PubChem CID 77124
IUPAC Name 3-N,3-N-dimethylbenzene-1,3-diamine;dihydrochloride
SMILES CN(C)C1=CC=CC(=C1)N.Cl.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 217.0°C
Color Purple to White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2NC6H4NH2·2HCl
Quantity 100 g
Beilstein 13, 40
Solubility Information Solubility in water: soluble
Formula Weight 209.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Dimethyl-m-phenylenediamine dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Dimethyl-m-phenylenediamine dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.