missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl Guanylmelamine Hemisulfate, TRC

Product Code. 30387590
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387590 25 mg 25mg
30383421 2.5 mg 2.5mg
30384647 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387590 Supplier Toronto Research Chemicals Supplier No. TRCD47374525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N-Dimethyl Guanylmelamine
Analyte or Component Names N,N-Dimethyl Guanylmelamine
Shipping Condition Room Temperature
Molecular Formula 2C6 H12 N8 . H2 O4 S
Synonym 1-(4-Amino-6-(dimethylamino)-1,3,5-triazin-2-yl)guanidine Hemisulfate, N,N-Dimethyl Metformin EP Impurity B Hemisulfate
InChI Formula InChI=1S/2C6H12N8.H2O4S/c2*1-14(2)6-12-4(9)11-5(13-6)10-3(7)8;1-5(2,3)4/h2*1-2H3,(H6,7,8,9,10,11,12,13);(H2,1,2,3,4)
Solution Type Neat
SMILES CN(C)c1nc(N)nc(NC(=N)N)n1.CN(C)c2nc(N)nc(NC(=N)N)n2.OS(=O)(=O)O
IUPAC Name 1-[4-amino-6-(dimethylamino)-1,3,5-triazin-2-yl]guanidine;sulfuric acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 490.5
Quantity 25 mg
Formula Weight 490.2044
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.