missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl Guanylmelamine Hemisulfate, TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyl Guanylmelamine |
| Analyte or Component Names | N,N-Dimethyl Guanylmelamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2C6 H12 N8 . H2 O4 S |
| Synonym | 1-(4-Amino-6-(dimethylamino)-1,3,5-triazin-2-yl)guanidine Hemisulfate, N,N-Dimethyl Metformin EP Impurity B Hemisulfate |
| InChI Formula | InChI=1S/2C6H12N8.H2O4S/c2*1-14(2)6-12-4(9)11-5(13-6)10-3(7)8;1-5(2,3)4/h2*1-2H3,(H6,7,8,9,10,11,12,13);(H2,1,2,3,4) |
| Solution Type | Neat |
| SMILES | CN(C)c1nc(N)nc(NC(=N)N)n1.CN(C)c2nc(N)nc(NC(=N)N)n2.OS(=O)(=O)O |
| IUPAC Name | 1-[4-amino-6-(dimethylamino)-1,3,5-triazin-2-yl]guanidine;sulfuric acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?