missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-D-glucamine Hydrochloride, TRC

Product Code. 30384149
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384149 10 g
30384733 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384149 Supplier Toronto Research Chemicals Supplier No. TRCD47348810G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 186541-49-1
Molecular Formula C8H19NO5 . HCl
Molecular Weight (g/mol) 245.7
Synonym 1-Deoxy-1-(dimethylamino)-D-glucitol Hydrochloride, N,N-Dimethylglucamine Hydrochloride
SMILES Cl.CN(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H19NO5.ClH/c1-9(2)3-5(11)7(13)8(14)6(12)4-10;/h5-8,10-14H,3-4H2,1-2H3;1H/t5-,6+,7+,8+;/m0./s1
Formula Weight 245.103
Percent Purity >95%
Chemical Name or Material N,N-Dimethyl-D-glucamine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.