missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitrosoaniline 99%, ACROS Organics™

Product Code. 10476720 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10476720 25 g Glass bottle 25g
10345750 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10476720 Supplier Thermo Scientific Acros Supplier No. 116490250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 138-89-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00002063
InChI Key CMEWLCATCRTSGF-UHFFFAOYSA-N
Synonym ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso
PubChem CID 8749
ChEBI CHEBI:59990
IUPAC Name N,N-dimethyl-4-nitrosoaniline
SMILES CN(C)C1=CC=C(C=C1)N=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83°C to 86°C
Color Green
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ONC6H4N(CH3)2
Quantity 25 g
Beilstein 12, 677
Fieser 01,746
Merck Index 15, 6725
Formula Weight 150.18
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Dimethyl-4-nitrosoaniline, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.