missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitrosoaniline 99%, ACROS Organics™

Product Code. 10345750 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10345750

Brand: Thermo Scientific Acros 116491000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 138-89-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00002063
InChI Key CMEWLCATCRTSGF-UHFFFAOYSA-N
Synonym ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso
PubChem CID 8749
ChEBI CHEBI:59990
IUPAC Name N,N-dimethyl-4-nitrosoaniline
SMILES CN(C)C1=CC=C(C=C1)N=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 83°C to 86°C
Color Green
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ONC6H4N(CH3)2
Quantity 100 g
Beilstein 12, 677
Fieser 01,746
Merck Index 15, 6725
Formula Weight 150.18
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material N, N-Dimethyl-4-nitrosoaniline, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.