missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitrosoaniline, 98%

Product Code. 11463847
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463847 100 g 100g
11453847 25 g 25g
11498770 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463847 Supplier Thermo Scientific Alfa Aesar Supplier No. A18301.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 138-89-6 | C8H10N2O | 150.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-89-6
Molecular Formula C8H10N2O
Molecular Weight (g/mol) 150.181
MDL Number MFCD00002063
InChI Key CMEWLCATCRTSGF-UHFFFAOYSA-N
Synonym ndma, accelerine, p-nitroso-n,n-dimethylaniline, n,n-dimethyl-p-nitrosoaniline, 4-nitroso-n,n-dimethylaniline, 4-nitrosodimethylaniline, p-nitrosodimethylanilide, paranitrosodimethylanilide, 4-dimethylamino nitrosobenzene, benzenamine, n,n-dimethyl-4-nitroso
PubChem CID 8749
ChEBI CHEBI:59990
IUPAC Name N,N-dimethyl-4-nitrosoaniline
SMILES CN(C)C1=CC=C(C=C1)N=O

Specifications

Melting Point 84°C to 87°C
Quantity 100 g
UN Number UN1369
Beilstein 607293
Merck Index 14,6638
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material N,N-Dimethyl-4-nitrosoaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.