missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4-nitroaniline, 98+%

Product Code. 11360024
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360024 5 g 5g
11370024 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360024 Supplier Thermo Scientific Alfa Aesar Supplier No. L00404.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-23-2 | C8H10N2O2 | 166.18 g/mol

It is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.

Solubility
Soluble in dichloromethane. Insoluble in water. Solubility in hot methanol (almost transparency).

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-23-2
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014712
InChI Key QJAIOCKFIORVFU-UHFFFAOYSA-N
Synonym 4-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-4-nitro, 4-nitrodimethylaniline, p-nitro-n,n-dimethylaniline, p-nitrodimethylaniline, 1-dimethylamino-4-nitrobenzene, 4-dimethylamino nitrobenzene, p-dimethylamino nitrobenzene, n,n-dimethyl-p-nitroaniline, aniline, n,n-dimethyl-p-nitro
PubChem CID 7491
IUPAC Name N,N-dimethyl-4-nitroaniline
SMILES CN(C)C1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 163°C to 166°C
Quantity 5 g
UN Number UN2811
Beilstein 638087
Solubility Information Soluble in dichloromethane. Insoluble in water. Solubility in hot methanol (almost transparency).
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material N,N-Dimethyl-4-nitroaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.