missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethyl-3-chloropropylamine Hydrochloride, Mikromol™
Description
N,N-Dimethyl-3-chloropropylamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N,N-Dimethyl-3-chloropropylamine Hydrochloride |
| CAS | 4-5-5407 |
| Analyte or Component Names | N,N-Dimethyl-3-chloropropylamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H12 Cl N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | N,N-Dimethyl-3-chloropropylamine Hydrochloride, 1-Chloropropyl(dimethyl)amine Hydrochloride, Benzydamine Hydrochloride Impurity G, Benzydamine Impurity G |
| InChI Formula | InChI=1S/C5H12ClN.ClH/c1-7(2)5-3-4-6;/h3-5H2,1-2H3;1H |
| SMILES | Cl.CN(C)CCCCl |
| IUPAC Name | 3-chloro-N,N-dimethylpropan-1-amine;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?