missing translation for 'onlineSavingsMsg'
Learn More

N,N’-Dimethyl-3,3’-dithiopropionamide, TRC

Product Code. 30380059
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380059 100 g
30383879 50 g
30380454 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380059 Supplier Toronto Research Chemicals Supplier No. TRCD472705100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-72-4
Molecular Formula C8 H16 N2 O2 S2
Molecular Weight (g/mol) 236.35
Synonym 3,3'-Dithiobis[N-methyl-propanamide, 3,3'-Dithiobis[N-methyl-propionamide, N,N'-Dimethyl-3,3'-dithiodipropionamide
IUPAC Name N-methyl-3-[[3-(methylamino)-3-oxopropyl]disulfanyl]propanamide
SMILES CNC(=O)CCSSCCC(=O)NC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H16N2O2S2/c1-9-7(11)3-5-13-14-6-4-8(12)10-2/h3-6H2,1-2H3,(H,9,11)(H,10,12)
Formula Weight 236.0653
Chemical Name or Material N,N'-Dimethyl-3,3'-dithiopropionamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.