missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1-naphthylamine, 99%

Product Code. 10266880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266880 25 mL Glass bottle 25mL
10688314 5 mL Glass bottle 5mL
10308440 100 mL Glass bottle 100mL
10723534 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10266880 Supplier Thermo Scientific Acros Supplier No. 204170250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-56-6 | C12H13N | 171.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-56-6
Molecular Formula C12H13N
Molecular Weight (g/mol) 171.24
MDL Number MFCD00003919
InChI Key AJUXDFHPVZQOGF-UHFFFAOYSA-N
Synonym n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin
PubChem CID 6848
IUPAC Name N,N-dimethylnaphthalen-1-amine
SMILES CN(C)C1=CC=CC2=CC=CC=C21

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.0400g/mL
Boiling Point 139°C to 140°C (13.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C10H7N(CH3)2
Refractive Index 1.6210 to 1.623
Quantity 25 mL
Beilstein 12, 1221
Merck Index 15, 3276
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 171.24
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyl-1-naphthylamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethyl-1-naphthylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.