missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-1-dodecylamine N-oxide, 95%

Product Code. 11332009
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332009 25 g 25g
11322009 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332009 Supplier Thermo Scientific Alfa Aesar Supplier No. H26065.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1643-20-5 | C14H31NO | 229.408 g/mol

N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyl-1-dodecylamine N-oxide is used as a foam builder and stabilizer. It is also used in cosmetics, emollients, conditioners, emulsifiers, dishwasher detergents, shampoos, antistatic, wetting, antibacterial and antifungal agents. Further, it is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1643-20-5
Molecular Formula C14H31NO
Molecular Weight (g/mol) 229.408
MDL Number MFCD00002049
InChI Key SYELZBGXAIXKHU-UHFFFAOYSA-N
Synonym lauramine oxide, lauryldimethylamine oxide, dodecyldimethylamine oxide, lauryldimethylamine n-oxide, ldao, ammonyx ao, ammonyx lo, empigen ob, aromox dmcd, conco xal
PubChem CID 15433
IUPAC Name N,N-dimethyldodecan-1-amine oxide
SMILES CCCCCCCCCCCC[N+](C)(C)[O-]

Especificaciones

Melting Point 120°C to 123°C
Density 0.996
Quantity 25 g
UN Number UN3259
Beilstein 1769927
Sensitivity Very Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 229.4
Percent Purity 95%
Chemical Name or Material N,N-Dimethyl-1-dodecylamine N-oxide
compliance-icons
Nombre del producto
  • N,N-Dimethyl-1-dodecylamine N-oxide
Palabra de advertencia
  • Danger
Clasificación
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Indicaciones de peligro
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Otros peligros
  • MIXTURE LIST-Contain: N,N-Dimethyl-1-dodecylamine N-oxide

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.