missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylenediamine, 97%

Product Code. 11381697
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381697 10 g 10g
11391697 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381697 Supplier Thermo Scientific Alfa Aesar Supplier No. L04765.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-05-1 | C8H20N2 | 144.262 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
N,N-Diisopropylethylenediamine can be used to produce 1,4-diisopropyl-piperazin-2-one by heating. It is used as ligands for metathesis catalysts. It acts as anti-microbial agent.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air, oxidizing agents. Incompatible materials are Strong oxidizing agents, Strong acids
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-05-1
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.262
MDL Number MFCD00015015
InChI Key CURJNMSGPBXOGK-UHFFFAOYSA-N
Synonym 2-aminoethyldiisopropylamine, n,n-diisopropylethylenediamine, 2-diisopropylamino ethylamine, usaf am-2, 1,2-ethanediamine, n,n-bis 1-methylethyl, n,n-diisopropyl-1,2-ethanediamine, n1,n1-diisopropylethane-1,2-diamine, 2-aminoethyl diisopropylamine, n,n-diisopropyl ethylenediamine, 2-diisopropylaminoethylamine
PubChem CID 8459
IUPAC Name N',N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)N(CCN)C(C)C

Specifications

Density 0.828
Boiling Point 170°C
Flash Point 54°C (129°F)
Refractive Index 1.442
Quantity 10 g
UN Number UN2733
Beilstein 1734800
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material N,N-Diisopropylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.