missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylethylenediamine, 97%

Product Code. 11428996
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428996 25 g 25g
11438996 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428996 Supplier Thermo Scientific Alfa Aesar Supplier No. A17114.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4013-94-9 | C8H20N2 | 144.26 g/mol

N,N'-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N′-Diethylethylenediamine is used in the preparation of 2-imidazolidinecarboxaldehydes and 1,4,6,9-tetraalkyl-1,4,6,9-tetraaza-5,10-dioxaperhydroanthracenes by reacting with glyoxal. It acts as a ligand and form coordination complexes with copper(II), which shows thermochromic properties.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4013-94-9
Molecular Formula C8H20N2
Molecular Weight (g/mol) 144.26
MDL Number MFCD00008864
InChI Key MFIGJRRHGZYPDD-UHFFFAOYSA-N
Synonym n,n'-diisopropylethylenediamine, 1,2-ethanediamine, n,n'-bis 1-methylethyl, 1,2-ethanediamine, n1,n2-bis 1-methylethyl, n,n'-diisopropylethylene diamine, n,n'-di propan-2-yl ethane-1,2-diamine, 1,2-bis isopropylamino ethane, n,n'-disopropylethylene diamine, 1,2-ethanediamine,n1,n2-dipropyl, diped, n,n'-diisopropyl-ethylenediamine
PubChem CID 77628
IUPAC Name N,N'-di(propan-2-yl)ethane-1,2-diamine
SMILES CC(C)NCCNC(C)C

Specifications

Density 0.798
Boiling Point 169°C to 171°C
Flash Point 51°C (123°F)
Refractive Index 1.4289
Quantity 25 g
UN Number UN2733
Beilstein 1735571
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 144.26
Percent Purity 97%
Chemical Name or Material N,N'-Diisopropylethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.