missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylethylamine (Peptide Synthesis), Fisher BioReagents

Product Code. 10647823
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647823 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10647823 Supplier Fisher BioReagents Supplier No. BP592500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
N,N-Diisopropylethylamine is commonly used in peptide synthesis reactions.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Diisopropylethylamine
CAS 7087-68-5
Melting Point -46°C
Color Yellow
Density 0.7560g/cm³
Boiling Point 127°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Molecular Formula C8H19N
Refractive Index 1.4113 to 1.4153 (at 25°C)
Quantity 500 mL
Merck Index 15, 3219
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
SMILES CCN(C(C)C)C(C)C
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
Molecular Weight (g/mol) 129.247
PubChem CID 81531
Formula Weight 129.24
Vapor Pressure 38mmHg at 31°C
Percent Purity ≥99%
Grade Peptide Synthesis
Physical Form Liquid
Chemical Name or Material N, N-Diisopropylethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Diisopropylethylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.