missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylcarbodiimide, TRC

Product Code. 30372583
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity unitSize
30372583 500 mg 500mg
1 options
This item is not returnable. View return policy

Product Code. 30372583

Brand: Toronto Research Chemicals TRCD455495500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Diisopropylcarbodiimide
CAS 693-13-0
Molecular Formula C7H14N2
Quantity 500 mg
Synonym N,N'-Methanetetraylbis-2-propanamine, 1,3-Diisopropylcarbodiimide, DIC, DIPC, DIPCDI, Diisopropylcarbodiimide, NSC 42080
InChI Formula InChI=1S/C7H14N2/c1-6(2)8-5-9-7(3)4/h6-7H,1-4H3
SMILES c1ccc(cc1)COc2ccc(cc2)[C@H]3C4(CCC(=O)CC4)C(=O)N3c5ccc(F)cc5
IUPAC Name N,N'-di(propan-2-yl)methanediimine
Molecular Weight (g/mol) 126.2
Formula Weight 126.1157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.