missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylcarbodiimide, 99%, AcroSeal™

Product Code. 11945591 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11945591 100 mL AcroSeal™ Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Product Code. 11945591 Supplier Thermo Scientific Acros Supplier No. 446181000
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 693-13-0 | C7H14N2 | 126.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 693-13-0
Density 0.8100g/mL
Boiling Point 145.0°C to 148.0°C
Flash Point 33°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Molecular Formula C7H14N2
Linear Formula (CH3)2CHN=C=NCH(CH3)2
Refractive Index 1.4320 to 1.4350
MDL Number MFCD00065689
Quantity 100 mL
Merck Index 15,3218
Specific Gravity 0.81
Synonym n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
Solubility Information Solubility in water: decomposes
InChI Key BDNKZNFMNDZQMI-UHFFFAOYSA-N
SMILES CC(C)N=C=NC(C)C
IUPAC Name N,N'-di(propan-2-yl)methanediimine
Molecular Weight (g/mol) 126.2
PubChem CID 12734
ChEBI CHEBI:53092
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material N, N'-Diisopropylcarbodiimide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N'-Diisopropylcarbodiimide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.