missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diisopropylcarbodiimide, 99%, Thermo Scientific™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass Bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
693-13-0
C7H14N2
126.2
MFCD00065689
BDNKZNFMNDZQMI-UHFFFAOYSA-N
n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
12734
CHEBI:53092
N,N'-di(propan-2-yl)methanediimine
CC(C)N=C=NC(C)C
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
693-13-0
C7H14N2
126.2
MFCD00065689
BDNKZNFMNDZQMI-UHFFFAOYSA-N
n,n'-diisopropylcarbodiimide, diisopropylcarbodiimide, 1,3-diisopropylcarbodiimide, n,n-diisopropylcarbodiimide, carbodiimide, diisopropyl, dipcdi, dic, 2-propanamine, n,n'-methanetetraylbis, diisopropylmethanediimine, n,n'-methanediylidenebis propan-2-amine
12734
CHEBI:53092
N,N'-di(propan-2-yl)methanediimine
CC(C)N=C=NC(C)C

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Coupling reagent
CAS Min % 98.5
CAS Max % 100.0
Density 0.8100g/mL
Color Colorless to Yellow
Boiling Point 145.0°C to 148.0°C
Flash Point 33°C
Assay Percent Range 98.5% min. (GC)
Infrared Spectrum Authentic
Linear Formula (CH3)2CHN=C=NCH(CH3)2
UN Number 3384
Packaging Glass Bottle
Merck Index 15,3218
Refractive Index 1.4320 to 1.4350
Quantity 100g
Solubility Information Solubility in water: decomposes
Specific Gravity 0.81
Formula Weight 126.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material N, N'-Diisopropylcarbodiimide, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N'-Diisopropylcarbodiimide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only