missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylaniline, 97%

Product Code. p-7052716
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491728 250 mg 250mg
15471728 1 g 1g
15481728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491728 Supplier Thermo Scientific Alfa Aesar Supplier No. H55471.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4107-98-6 | C12H19N | 177.291 g/mol

N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.

Notes
Light Sensitive. Avoid oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4107-98-6
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00048277
InChI Key OVSARSKQWCLSJT-UHFFFAOYSA-N
Synonym n,n-diisopropylaniline, benzenamine, n,n-bis 1-methylethyl, unii-kep9pka41k, kep9pka41k, n,n-dsopropylanlne, diisopropylphenyl amine, acmc-1asew, dsstox_cid_22189, dsstox_rid_79949, dsstox_gsid_42189
PubChem CID 61329
IUPAC Name N,N-di(propan-2-yl)aniline
SMILES CC(C)N(C1=CC=CC=C1)C(C)C

Specifications

Color Colorless to Yellow
Density 0.91 g/mL
Boiling Point 95°C to 96°C (11 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.519
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 177.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N,N-Diisopropylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.