missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylaniline, 97%

Product Code. 15471728
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15471728

Brand: Thermo Scientific Alfa Aesar H55471.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4107-98-6 | C12H19N | 177.291 g/mol

N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diisopropylaniline was used for the synthesis of 4-diisopropylamino benzonitrile. It may be used for the synthesis of 6-(4-bromophenyl)-3-methoxy-5-methyl-8-oxabicyclo[3.2.1]octa-3,6-dien-2-one.

Notes
Light Sensitive. Avoid oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4107-98-6
Molecular Formula C12H19N
Molecular Weight (g/mol) 177.291
MDL Number MFCD00048277
InChI Key OVSARSKQWCLSJT-UHFFFAOYSA-N
Synonym n,n-diisopropylaniline, benzenamine, n,n-bis 1-methylethyl, unii-kep9pka41k, kep9pka41k, n,n-dsopropylanlne, diisopropylphenyl amine, acmc-1asew, dsstox_cid_22189, dsstox_rid_79949, dsstox_gsid_42189
PubChem CID 61329
IUPAC Name N,N-di(propan-2-yl)aniline
SMILES CC(C)N(C1=CC=CC=C1)C(C)C

Specifications

Color Colorless to Yellow
Density 0.91 g/mL
Boiling Point 95°C to 96°C (11 mmHg)
Flash Point 84°C (183°F)
Refractive Index 1.519
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 177.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N,N-Diisopropylaniline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.