missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diethylthiourea, 98%

Product Code. 11413367
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413367 250 g 250g
11403367 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413367 Supplier Thermo Scientific Alfa Aesar Supplier No. A17143.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 105-55-5 | C5H12N2S | 132.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-55-5
Molecular Formula C5H12N2S
Molecular Weight (g/mol) 132.23
MDL Number MFCD00004925
InChI Key FLVIGYVXZHLUHP-UHFFFAOYSA-N
Synonym 1,3-diethyl-2-thiourea, n,n'-diethylthiourea, pennzone e, thiourea, n,n'-diethyl, thiate h, n,n'-diethylthiocarbamide, urea, 1,3-diethyl-2-thio, usaf ek-1803, n,n-diethyl-2-thiourea, diethylthiourea
PubChem CID 2735009
ChEBI CHEBI:82448
IUPAC Name 1,3-diethylthiourea
SMILES CCNC(=S)NCC

Specifications

Melting Point 75°C to 79°C
Density 1.12
Flash Point 186°C (367°F)
Quantity 250 g
Beilstein 773905
Formula Weight 132.23
Percent Purity 98%
Chemical Name or Material N,N'-Diethylthiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.