missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylsalicylamide, 97%

Product Code. 15471138
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471138 1 g 1g
15481138 5 g 5g
15491138 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471138 Supplier Thermo Scientific Alfa Aesar Supplier No. H55025.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19311-91-2 | C11H15NO2 | 193.246 g/mol

N,N-Diethylsalicylamide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethylsalicylamide is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19311-91-2
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00010816
InChI Key ZVYXEXAXXWINEH-UHFFFAOYSA-N
Synonym n,n-diethylsalicylamide, diethylsalicylamide, salicyldiethylamide, salicylamide, n,n-diethyl, benzamide, n,n-diethyl-2-hydroxy, o-hydroxy-n,n-diethylbenzamide, unii-hg05mo88tl, hg05mo88tl, acmc-20ak91, 2-hydroxy-n,n-diethylbenzamide
PubChem CID 88006
IUPAC Name N,N-diethyl-2-hydroxybenzamide
SMILES CCN(CC)C(=O)C1=CC=CC=C1O

Specifications

Melting Point 93°C to 97°C
Quantity 1 g
Formula Weight 193.25
Percent Purity 97%
Chemical Name or Material N,N-Diethylsalicylamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.