missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylpropargylamine, 97%

Product Code. 15851012 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15851012 100 g 100g
15831012 5 g 5g
15841012 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15851012 Supplier Thermo Scientific Acros Supplier No. 455581000
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 4079-68-9 | C7H13N | 111.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4079-68-9
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.18
InChI Key JZJXKEWVUBVOEH-UHFFFAOYSA-N
PubChem CID 20010

Specifications

Density 0.8110g/mL
Boiling Point 118°C to 120°C
Flash Point 20°C
Assay Percent Range 96% min. (GC)
Quantity 100 g
Specific Gravity 0.811
Formula Weight 111.18
Percent Purity 97%
Chemical Name or Material N,N-Diethylpropargylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.