missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylnicotinamide, 97%

Product Code. 10288292 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10288292 25 g Glass bottle 25g
10625152 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10288292 Supplier Thermo Scientific Supplier No. 114470250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-26-7 | C10H14N2O | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-26-7
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.23
MDL Number MFCD00006386
InChI Key NCYVXEGFNDZQCU-UHFFFAOYSA-N
Synonym nikethamide, n,n-diethylnicotinamide, nicethamide, corvin, anacardone, anacordone, carbamidal, coramine, corvotone, nicamide
PubChem CID 5497
IUPAC Name N,N-diethylpyridine-3-carboxamide
SMILES CCN(CC)C(=O)C1=CN=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 296°C to 300°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.526
Quantity 25 g
Beilstein 22, II, 34
Merck Index 15, 6627
Specific Gravity 1.06
Solubility Information Solubility in water: miscible. Other solubilities: miscible with methanol,ethanol,acetone,chlorofo,rm,ether
Formula Weight 178.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Diethylnicotinamide, 97%
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • N,N-Diethylnicotinamide
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.